Order Entry
United States
ContactUsLinkComponent
 
Fluorescein Labeled Lycopersicon Esculentum Lectin (LEL), Vector Laboratories
Catalog # 101098-006
CAS Number:  
undefined
Fluorescein Labeled Lycopersicon Esculentum Lectin (LEL), Vector Laboratories
Catalog # 101098-006
Supplier Number:  FL-1171-1
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Host:
  • Description:
    Fluorescein Labeled Lycopersicon Esculentum Lectin, Green
  • Size:
    1 mg
  • Environmentally Preferable:
  • Protein:
    Lycopersicon Esculentum Lectin, 1 mg
  • Cat. No.:
    101098-006
  • Conjugate:
    Fluorescein
  • Supplier no.:
    FL-1171

Specifications

About this item

Fluorescein labeled Lycopersicon Esculentum (Tomato) Lectin (LEL, TL). Tomato lectin is a very stable single subunit glycoprotein containing about 50 percent arabinose and galactose and may form multimeric aggregates in solution. LEL binds well to glycophorin and Tamm-Horsfall glycoprotein and has been used effectively to label vascular endothelium in rodents.

  • Excitation maximum : 495 nm
  • Emission maximum : 515 nm
  • Inhibiting/eluting sugar : chitin hydrolysate

Tomato lectin (from Lycopersicon esculentum) is an effective marker of blood vessels and microglial cells in rodents

Accompanying each fluorescent lectin is an analysis data sheet summarizing the results of our quality control tests and providing pertinent information on the product. All of these reagents are supplied as solutions perserved with sodium azide.

Fluorescein labeled Tomato lectin has an appropriate number of fluorochromes bound to provide the optimum staining characteristics for this lectin. This conjugate is supplied essentially free of unconjugated fluorochromes.

Conjugation of the lectin with a fluorophore facilitates fast, one-step detection and visualization using intravascular perfusion methods or direct application to tissue sections.