Order Entry
United States
ContactUsLinkComponent
Chocolate Agar, Plates, Northeast Lab Services
Chocolate Agar, Plates, Northeast Lab Services
Catalog # 101373-946
CAS Number:  
Chocolate Agar, Plates, Northeast Lab Services
Catalog # 101373-946
Supplier Number:  P1150
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Media Format:
    Liquid
  • Description:
    Chocolate Agar with Enrichment
  • Size:
    15 mm x 100 mm
  • Environmentally Preferable:
  • Cat. No.:
    101373-946
  • Fill volume:
    20 ml
  • Supplier no.:
    P1150

Specifications

About this item

Chocolate Agar is contained in a standard petri dish.

  • Standard Petri Dish
  • Fill Volume: 20 mL
  • Size: 15 mm x 100 mm
  • Prepared Medium
  • Enrichment Included

The petri dish measures 15 mm x 100 mm, and the fill volume is 20 mL.

NEL manufactures over 500 proprietary products and over 1300 classic media formulations for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.

Certifications: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.