About this item
Endotoxin-free glass vials sealed individually with tear-out aluminum ring that recommended for storing and handling samples with special requirement of endotoxin level (<0.05 EU/ml).
- Keeps endotoxin away from samples and products
- Recommended for storing and handling samples with special requirement of endotoxin level
- Endotoxin Free Glass Vials
Endotoxin is the lipopolysaccharide (LPS) complex located in the outer membrane of gram-negative bacteria. During experimental procedures, large amount of endotoxins are shed and can easily contaminate labware, buffers and downstream products. Presence of endotoxin in any of these has a detrimental effect on culturing of cells and can lead to generation of irreproducible or negative experimental data. Therefore, it is critical to keep endotoxin away from samples and products. ToxOut™ Endotoxin-free Glass Vials are sealed individually with tear-out aluminum ring and are recommended for storing and handling samples with special requirement of endotoxin level (<0.05 EU/ml).