ExoPure™ 1.0 µm Immunobeads, Overall Exosome Isolation, Plasma, Urine, Serum, BioVision Inc
Catalog # 75799-352
Supplier: Avantor
CAS Number:
Specifications
- Description:1 micron pre-coupled latex beads (Exosome Isolation, plasma, urine, serum)
- Size:10 reactions
- Environmentally Preferable:
- Cat. No.:75799-352
- Supplier no.:M1039-10
Specifications
About this item
1.0 micron Immunobeads for overall exosome isolation from human biofluids (plasma, serum, urine).
- 1.0 micron Immunobeads for overall exosome isolation from human biofluids (tested for plasma, serum, urine)
- Small sample volume of biofluids
- No ultracentrifugation or other methods for exosome purification required
- Ready to use and stable for long term (up to 8 months)
- Suitable for nucleic acid extraction, protein profiling of exosome markers, exosome elution from immunobeads
- A mouse monoclonal antibody has been used for coating