ContactUsLinkComponent
BBL™ CHROMagar™ Salmonella, BD
BBL™ CHROMagar™ Salmonella, BD
  90006-158
 :  Avantor
 :  
BBL™ CHROMagar™ Salmonella, BD
  90006-158
 :  Avantor
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    BBL™ CHROMagar™ Salmonella, AOAC
  • Size:
    100 mm
  • Environmentally Preferable:
  • Cat. No.:
    90006-158
  • Supplier no.:
    214983
BBL™ CHROMagar™ Salmonella is a selective and differential medium for the isolation and presumptive identification of Salmonella species.
  • Cost-effective
  • Detects Salmonella in as little as 24 hours
  • Salmonella species produce mauve-colored colonies
BBL™ CHROMagar™ Salmonella is a selective and differential medium for the isolation and presumptive identification of Salmonella species from other coliform and non-coliform bacteria in clinical stool samples and a variety of food samples.