ContactUsLinkComponent
Sabouraud Dextrose Agar, BD
Sabouraud Dextrose Agar, BD
Catalog # 90005-224
Supplier:  BD
CAS Number:  
Sabouraud Dextrose Agar, BD
Catalog # 90005-224
Supplier:  BD
Supplier Number:  299510
CAS Number:  
Specifications
  • Description:
    Sabouraud Dextrose Agar, Sterile Pack
  • Size:
    500 mL
  • Environmentally Preferable:
  • Cat. No.:
    90005-224
  • Supplier no.:
    299510
Specifications
About this item
Sabouraud Dextrose Agar is used for the cultivation of dermatophytes.
  • Peptones provide a source of nitrogen
  • Dextrose is an energy source
  • Agar is the solidifying agent
After sufficient incubation, the plates should show isolated colonies in streaked areas and confluent growth in areas of heavy inoculation.