ContactUsLinkComponent
BD BBL™ CHROMagar™ Candida, BD Diagnostics
BD BBL™ CHROMagar™ Candida, BD Diagnostics
Catalog # 90003-480
Supplier:  BD
CAS Number:  
undefined
BD BBL™ CHROMagar™ Candida, BD Diagnostics
Catalog # 90003-480
Supplier:  BD
Supplier Number:  254093
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    BBL™ CHROMagar™ Candida
  • Size:
    100 mm
  • Environmentally Preferable:
  • Cat. No.:
    90003-480
  • Supplier no.:
    254093
Specifications
About this item
BBL™ CHROMagar™ Candida is a selective medium for the isolation and presumptive identification of yeast and filamentous fungi and differentiation of Candida albicans, C
  • Peptones supply essential nutrients
  • Chloramphenicol inhibits most bacterial contaminants
  • Agar is the solidifying agent
Due to the differences in morphology and colors of the yeast colonies, this medium facilitates the detection of mixed yeast cultures in specimens. It may also be used as a selective isolation medium for other yeasts and for filamentous fungi instead of Sabouraud Dextrose Agar or similar media.
tropicalis and C. krusei.