Order Entry
United States
ContactUsLinkComponent
Transglutaminase Activity Assay Kit (Colorimetric), BioVision
Transglutaminase Activity Assay Kit (Colorimetric), BioVision
Catalog # 75836-744
Supplier:  Avantor
CAS Number:  
undefined
Transglutaminase Activity Assay Kit (Colorimetric), BioVision
Catalog # 75836-744
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Transglutaminase Activity Assay Kit (Colorimetric)
  • Size:
    100 Assays
  • Environmentally Preferable:
  • Detection Range:
    ~10 µU or 80 ng of recombinant hTG2 enzyme
  • Cat. No.:
    75836-744
  • Supplier no.:
    K571-100

Specifications

About this item

Validated using many sample types.

  • Simple, rapid & convenient
  • The assay can detect transglutaminase activity ~10 µU or 80 ng of recombinant hTG2 enzyme
  • Detection method- Absorbance (525 nm)
  • Applications- Measurement of Transglutaminase activity

Transglutaminases (EC 2.3.2.13) are calcium dependent enzymes that catalyze the post-translational modification of proteins by formation of isopeptide bonds. This occurs either through protein cross-linking via formation of γ-glutamyl-ε-lysine bonds or through incorporation of primary amines at selected peptide-bound glutamine residues. The transglutaminase enzyme family comprises the intracellular forms (TG1, TG3 and TG5) expressed mostly in the epithelial tissue; TG2 which is both intracellular and extracellular and expressed in various tissue types; TG4 which is expressed in the prostate gland; factor XIII which is expressed in blood; TG6 and TG7, whose tissue distribution is unknown and band 4.2 (lacking enzymatic activity) which is present on erythrocyte membranes. Transglutaminases also exhibit GTPase, phosphodiesterase and protein kinase activity. Transglutaminases are associated with certain neurological and autoimmune disorders and also cancer. BioVision’s Transglutaminase Activity Assay kit utilizes the deamidation reaction of the transglutaminase enzyme with a donor and acceptor substrate resulting in the formation of a hydroxamate product. The hydroxamate product reacts with the Stop Solution forming a purple complex that can be measured colorimetrically at 525 nm. The limit of quantification of this assay is ~10 µU or 80 ng of recombinant hTG2 enzyme.