- Description:Mannitol YEP (Yeast Extract Peptone) Agar
- Size:Pack of 10
- Environmentally Preferable:
- pH:6.2
- Minimum Order Quantity:10
- Cat. No.:10789-386
- Volume:20 ml
- Supplier no.:G39
For use in the cultivation and enumeration of microorganisms and growth promotion testing for aloe products.
For use in the cultivation and enumeration of microorganisms and growth promotion testing for aloe products.