Order Entry
United States
ContactUsLinkComponent
HISCRIBE™ T7 ARCA mRNA Kit, New England Biolabs
HISCRIBE™ T7 ARCA mRNA Kit, New England Biolabs
Catalog # 102902-442
Small Business Enterprise
CAS Number:  
undefined
HISCRIBE™ T7 ARCA mRNA Kit, New England Biolabs
Catalog # 102902-442
Supplier Number:  E2065S
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Description:
    HiScribe T7 ARCA mRNA Kit
  • Size:
    20 Reactions
  • Environmentally Preferable:
  • Cat. No.:
    102902-442

Specifications

About this item

Most eukaryotic mRNAs require a 7-methyl guanosine (m7G) cap structure at the 5´ end and a Poly(A) tail at the 3´ end to be efficiently translated.

By using a DNA template encoding a poly(A) tail, the HiScribe T7 ARCA mRNA Kit can be used to synthesize capped and tailed mRNAs. The cap structure is added to the mRNA by co-transcriptional incorporation of Anti-Reverse Cap Analog (ARCA) using T7 RNA Polymerase. The transcription reaction can be set up easily by combining the ARCA/NTP Mix, T7 RNA Polymerase Mix and a suitable DNA template. The kit also allows for partial incorporation of 5mCTP, Pseudo-UTP and other modified nucleotides into mRNA. mRNAs synthesized with the kit can be used for cell transfection, microinjection, in vitro translation and RNA vaccines. Poly(A) tail is incorporated during the transcription reaction. The kit also includes DNase I and LiCl for DNA template removal and quick mRNA purification.