About this item
Combines the components of the Alcian Blue and Periodic Acid Schiff (PAS) stain kits.
- pH 2.5
- For identifying acids
Alcian Blue is normally prepared at an acidic pH of 2.5 and is used to identify acid mucopolysaccharides and acetic mucins.
Excessive amounts of non-sulfated acidic mucosubstances are seen in mesotheliomas, certain amounts occur normally in blood vessel walls but increase in early lesions of atherosclerosis.