About this item
MM 24 is a low viscosity mounting medium that is compatible with both aliphatic (e.g. Xylene substitutes) and aromatic (e.g. xylene) clearing agents.
- Compatible with aliphatic and aromatic clearing agents
- Specially formulated
- Available in three sizes
It is specially formulated to provide optimum results with automatic glass coverslippers, yet is also effective when manually preparing slides
Available in Pint, 4/1 Pint Case, or 12/4oz. Case.
Set up time before permanent filing can be acieved as quickly as 24 hours when used with aromatic agents and 48 to 72 hours when used with aliphatics.