ContactUsLinkComponent
Difco™ Brilliant Green Agar, BD Life Sciences
  90002-000
 :  BD
 :  
Difco™ Brilliant Green Agar, BD Life Sciences
  90002-000
 :  BD
 :  228530
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Difco™ Brilliant Green Agar
  • Size:
    500 g
  • Environmentally Preferable:
  • Cat. No.:
    90002-000
  • Supplier no.:
    228530
Brilliant Green Agar is a highly selective medium for the isolation of Salmonella other than S
  • Brilliant green dye inhibits gram-positive bacteria
  • Inhibits most gram-negative bacilli
  • Phenol red serves as a pH indicator
Yellow coloring appears as a result of acid production in the fermentation of lactose and/or sucrose in the medium.
Typhi from feces and other materials.