Order Entry
United States
ContactUsLinkComponent
Tissue-Tek VIP®6 AI Vacuum Infiltration Processor, Sakura Finetek
Tissue-Tek VIP®6 AI Vacuum Infiltration Processor, Sakura Finetek
Catalog # 75818-110
Supplier:  Sakura Finetek
undefined
Tissue-Tek VIP®6 AI Vacuum Infiltration Processor, Sakura Finetek
Catalog # 75818-110
Supplier:  Sakura Finetek
Supplier Number:  6040
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Tissue-Tek VIP®6 AI Vacuum Infiltration Processor
  • Environmentally Preferable:
  • Cat. No.:
    75818-110
  • Electrical:
    115 VAC, 50/60 Hz, 12 A

Specifications

About this item

The first and only tissue procesor offering automatic, onboard preparation of mixed reagent solutions for advanced infiltration of fatty tissues.

  • Automatic Transfer allows in-process solution rotation
  • Automatic paraffin rotation and waste container save time and prevent spills
  • 2 bulk reagent reservoirs provide fresh reagent during processing, if needed
  • Heated retort lid reduces condensation while eliminating fumes
  • Automatic bottle connection check to reduce user errors
  • User-defined cleaning cycles w/optional drying cycle to eliminate fumes
  • Illuminated reagent cabinet enables observation of reagent levels
  • Automatic paraffin degassing aspirates evaporated clearing agent from paraffin oven, keeping paraffin clean while filtering the fumes

Most reliable processing with industry leading mean time between repairs of over 52 weeks. The high-resolution LCD color touch screen and user-friendly software is capable of up to 300 cassettes per run. Options for processing include 10 user-selectable mixing modes, heat, pressure and vacuum. Tissue-Tek® iSupport™ remote system monitorying Sakura's professional technical support team ensure maximum uptime.

Certifications: UL61010-1;2nd Ed.:2004
CAN/CSA-C22.2 No.61010-1-12
IEC 61010-2-010:2003
IEC 61010-2-101:2002.
Accessories information: Accessories include two activated carbon fume filters, two 150-cassette baskets, one USB memory stick, one set of reagent bottle labels, once case of paraffin waste bags, one large paraffin scraper, one small paraffin scraper, one basket transport tray, one level sensor cleaning brush and one operating manual.