Order Entry
United States
ContactUsLinkComponent
Beta Galactosidase (β-Gal) Inhibitor Screening Kit (Fluorometric), BioVision Inc.
Beta Galactosidase (β-Gal) Inhibitor Screening Kit (Fluorometric), BioVision Inc.
Catalog # 75832-884
Supplier:  Avantor
CAS Number:  
undefined
Beta Galactosidase (β-Gal) Inhibitor Screening Kit (Fluorometric), BioVision Inc.
Catalog # 75832-884
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Beta Galactosidase (β-Gal) Inhibitor Screening Kit (Fluorometric)
  • Size:
    100 assays
  • Environmentally Preferable:
  • Sample Type:
    Variable: Inhibitors, small molecules, proteins, etc.
  • Shelf Life:
    12 Months
  • Ex/em:
    480/520 nm
  • Cat. No.:
    75832-884
  • Storage temperature:
    –20 °C
  • Supplier no.:
    K827-100

Specifications

About this item

Kit for screening beta galactosidase Inhibitors.

  • Simple, HTP compatible, 30 minutes protocol
  • A simple and high-throughput adaptable assay kit
  • Used to fluorometrically screen, study and characterize potential inhibitors of beta galactosidase within 30 minutes

Beta Galactosidase (β-Gal, EC: 3.2.1.23) is an enzyme which hydrolyzes the β-galactosides into monosacchanides. β-Gal is widely used as a reporter gene in the field of molecular biology. Senescence Associated β-Gal (SA-β-Gal) is an isoform of β-Gal which has the optimal activity at pH 6.0, and is mostly used as a biomarker for senescent cells (K802). β-Gal is an essential enzyme in humans and its deficiency results in Morquio’s Syndrome, a severe birth defect. β-Gal can also be used as a tool to study protein-protein interaction. In BioVision’s Beta-Galactocidase Inhibitor Screening Kit, β-Gal converts β-Gal substrate to give an intensely fluorescent product. In the presence of a β-Gal inhibitor, the reaction is impeded/abolished resulting in decrease or total loss of fluorescence. This assay kit can be used to screen/study/characterize the potential inhibitors of Beta Galactosidase. The assay is simple, high-throughput adaptable and can be performed within 30 minutes.