To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Dimethylated Histone H3 Lysine 4, H3K4me2
- Antigen Symbol:H3K4me2
- Clonality:Monoclonal
- Clone:Clone: RM135
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Multiple
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Epitope:Dimethyllysine 4 in Histone H3
- Format:Purified
- Form:Liquid
- Gene ID:8350
- Storage Buffer:50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
- Storage Temperature:-20.0°C
- Concentration:1 mg/mL
- Shipping Temperature:4°C
- Immunogen:A dimethyl-peptide corresponding to Dimethyl-Histone H3 (Lys4)
- Purification:Protein A affinity purified from an animal origin-free culture supernatant
- Cat. No.:103255-446
- Supplier no.:31-1037-00
Specifications
About this item
This antibody reacts to Histone H3 dimethylated at Lysine 4 (K4me2). No cross reactivity with monomethylated Lysine 4 (K4me1) or trimethylated Lysine 4 (K4me3), or other methylation in histone H3.
Western Blot: 0.02 ug/mL – 0.2 ug/mL;
ICC: 1 ug/mL - 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.