ContactUsLinkComponent
Difco™ Tomato Juice Agar, BD Diagnostics
Difco™ Tomato Juice Agar, BD Diagnostics
Catalog # 90000-358
Supplier:  BD
CAS Number:  
Difco™ Tomato Juice Agar, BD Diagnostics
Catalog # 90000-358
Supplier:  BD
Supplier Number:  211794
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Difco™ Tomato Juice Agar
  • Size:
    500 g
  • Environmentally Preferable:
  • Cat. No.:
    90000-358
  • Supplier no.:
    211794
Specifications
About this item
Tomato Juice Agar is used for cultivating and enumerating Lactobacillus species.
  • Tomato juice is a source of carbon, protein, and nutrients
  • Peptone provides a source of nitrogen, amino acids, and carbon
  • Peptonized milk contains lactose as an energy source
  • Agar is the solidifying agent
Tomato Juice Agar is especially useful in cultivating L
Tomato Juice Agar, Special is recommended for the direct plate count of lactobacilli from saliva and for cultivation of other acidophilic microorganisms. This medium is more selective for lactobacilli than Tomato Juice Agar.
acidophilus from clinical specimens and foodstuffs.