Anti-KIR2DL1 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 143211]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: KIR2DL1
Clonality: Monoclonal
Clone: 143211
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103251-866
- 103251-864
- Antigen Symbol:
- KIR2DL1
- KIR2DL1
- Antigen Synonyms:
- KIR2DS5
- KIR2DS5
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- 143211
- 143211
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Immunogen:
- BaF3 mouse pro-B cell line transfected with human KIR2DL1/KIR2DS5 Accession # P43626
- BaF3 mouse pro-B cell line transfected with human KIR2DL1/KIR2DS5 Accession # P43626
- Purification:
- Protein A or G purified from hybridoma culture supernatant
- Protein A or G purified from hybridoma culture supernatant
- Storage Buffer:
- Supplied in a saline solution containing BSA and Sodium Azide.
- Supplied in a saline solution containing BSA and Sodium Azide.
- Storage Temperature:
- Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
- Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Specifications
Frequently Bought Together