Order Entry
United States
ContactUsLinkComponent
Anti-SSEA-4 Mouse Monoclonal Antibody (Fluorescein) [clone: MC-813-70]
Anti-SSEA-4 Mouse Monoclonal Antibody (Fluorescein) [clone: MC-813-70]
Supplier:  Novus Biologicals
Certificates
Anti-SSEA-4 Mouse Monoclonal Antibody (Fluorescein) [clone: MC-813-70]
Supplier:  Novus Biologicals
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 103244-648
  • 103244-650
  • Antigen Symbol:
  • SSEA4
  • SSEA4
  • Antigen Name:
  • Stage-specific Embryonic Antigen 4
  • Stage-specific Embryonic Antigen 4
  • Antigen Synonyms:
  • SSEA-4
  • SSEA-4
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Fluorescein
  • Fluorescein
  • Clone:
  • MC-813-70
  • MC-813-70
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG3
  • IgG3
  • Immunogen:
  • 2120Ep human embryonal carcinoma cell line
  • 2120Ep human embryonal carcinoma cell line
  • Purification:
  • Protein A or G purified from hybridoma culture supernatant
  • Protein A or G purified from hybridoma culture supernatant
  • Storage Buffer:
  • Supplied in a saline solution containing BSA and Sodium Azide.
  • Supplied in a saline solution containing BSA and Sodium Azide.
  • Storage Temperature:
  • Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
  • Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.

Specifications

Frequently Bought Together