Human Recombinant Dicer Enzyme
Catalog # 10497-944
Supplier: Avantor
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:
- Species:Human
- Size:4×50 units
- Environmentally Preferable:
- Enzyme Name:Dicer Enzyme
- Cat. No.:10497-944
- Supplier no.:T510008
Specifications
About this item
The Recombinant Human Dicer Enzyme Kit uses an ultra-active form of human recombinant dicer enzyme that can cleave more than 95% of dsRNA template into 22 bp siRNAs within 12 hours under optimized reaction conditions
- A mixture of siRNAs has a high chance of success than a single siRNA design
- More regions of the genes can be screened for silencing
- Stable at -20 °C for six months
This provides sufficient siRNAs in every single species to achieve effective gene silencing.