PM i™ + XY500 Filling System, bioMerieux
Supplier: Avantor
Certificates
About this item
Automated filling systems for tubes, flasks, and bottles. The filling system consists of a high precision dispenser and a highly repeatable robotic module.
- Automated filling of tubes, bottles, and flasks for enhanced lab productivity
- Compatible with XY500 and Masterclave for dispensing media
- Two racks 252 tubes each can be filled in the same cycle using dynamic loading from original packaging
- 4 different 96 tube rack configurations
- Switch from tube to bottle fillings in seconds
- Program the XY500 with specific settings in less than 1 min
- Fill over 1900 tubes/hour
Designed for the automated dispensing of broth or agar into plates, tubes, or bottles
Standardize dispensing with:
Accuracy and repeatability compliant with ISO 7218
Automatic traceability
Customizable labels
Bidirectional connectivity with LIMS communication
Experience total ease-of-use:
Smart software, color changing LEDs inform user of system status
RFID capability
Intuitive interface
Program management
Manual and automatic modes
Improve productivity:
1.6 L/min. (225 mL under 10 sec.; 500 mL under 20 sec.)
Certifications: The PMi Peristaltic Pump has accuracy and repetability compliant with ISO 7218.
Ordering information: The pump is delivered with 1x3.2 mm tubing set with nozzles, 1 foot switch, 1 power supply cable (90 W), 1 certificate of conformance, 1 quick start guide and 1 user manual (CD-ROM) and LabelManager for label customization.
Frequently Bought Together