Anti-RSK1/RSK2 Rabbit Polyclonal Antibody
Catalog # 103223-898
Supplier: Novus Biologicals
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:RSK1/RSK2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 ug
- Environmentally Preferable:
- Modification:p Ser221, p Ser227
- Storage Buffer:Supplied as a 0.2 μm filtered solution in PBS with Sodium Azide. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C, as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 6 months, -20 to -70 °C under sterile conditions after opening.
- Immunogen:Phosphopeptide containing human RSK1 S221 site
- Purification:Immunogen affinity purified
- Cat. No.:103223-898
- Supplier no.:AF892
Specifications
About this item
Type: Primary Antibodies
Antigen:
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat