ContactUsLinkComponent
VWR® Replacement Fiberglass Sleeve for Beaker Tongs
VWR® Replacement Fiberglass Sleeve for Beaker Tongs
Catalog # 10806-314
Supplier:  VWR International
undefined
VWR® Replacement Fiberglass Sleeve for Beaker Tongs
Catalog # 10806-314
Supplier:  VWR International
Specifications
  • Description:
    VWR® Replacement Fiberglass Sleeve for Beaker Tongs
  • Length:
    152 mm (6")
  • Environmentally Preferable:
  • Cat. No.:
    10806-314
Specifications
About this item
Replacement fiberglass sleeve covered jaws are designed for use with beaker tongs.
  • Fiberglass construction
  • Passivated
  • Autoclavable
They are passivated, and compatible with Gamma/ETO/Autoclavable sterilization methods.