Attest™ Rapid 5 Steam Plus Test Pack, 3M™
Supplier: Healthcare Products
Certificates
About this item
Fast, easy to use steam test packs contain a three hour Attest™ Rapid Readout Biological Indicator (BI) plus a moving front (SteriGage™) Class 5 Integrating Indicator.
Frequently Bought Together