TransIT®-293 Transfection Reagent, Mirus Bio
Supplier: Mirus Bio
Certificates
About this item
A high-efficiency, low-toxicity DNA transfection reagent optimized for 293 cells.
- Unique formulation
- High-efficiency delivery
- Deliver single or multiple plasmids
- Low cellular toxicity
Maximize transfection performance in HEK 293 cells with this reagent's unique formulation, and achieve 75-85% transfection efficiency to ensure experimental success.
Specifications
- Environmentally Preferable:
Specifications