ContactUsLinkComponent
Metal Backed Student Thermometer, United Scientific Supplies
Metal Backed Student Thermometer, United Scientific Supplies
Catalog # 470222-092
undefined
Metal Backed Student Thermometer, United Scientific Supplies
Catalog # 470222-092
Supplier Number:  THMC02
Specifications
  • Environmentally Preferable:
  • Cat. No.:
    470222-092
  • Range:
    −40…+110 °C
  • Supplier no.:
    THMC02
Specifications
About this item
Great for classroom use.
  • Glass thermometer mounted on aluminum back
  • Filled with kerosene solution
  • Celsius only
Glass thermometer mounted on aluminum back. Thermometers are filled with approximately 3 grams of kerosene mixed with red color.