Anti-CDK9 Mouse Monoclonal Antibody [clone: 1B5A7]
10168-244
: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Cyclin-dependent Kinase 9
- Antigen Symbol:CDK9
- Clonality:Monoclonal
- Clone:1B5A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:1025
- Antigen Synonyms:CDC2-related kinase|C-2k|tat-associated kinase complex catalytic subunit|CTK1|serine/threonine-protein kinase PITALRE|Cell Division Cycle 2-like protein kinase 4|cell division protein kinase 9|PITALRE|TAK|serine/threonine protein kinase PITALRE|CDC2L4|RP11-228B15.5
- Storage Buffer:Ascitic fluid containing 0.03% sodium azide.
- Storage Temperature:4 °C; –20 °C for long term storage
- Immunogen:The immunogen corresponding to human CDK9 expressed in E. Coli.
- Cat. No.:10168-244
- Supplier no.:GTX60442
Mouse Monoclonal Antibody [1B5A7] to CDK9
Type: Primary
Antigen: CDK9
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human