Order Entry
United States
ContactUsLinkComponent
Anti-SHANK3 Rabbit Polyclonal Antibody
Anti-SHANK3 Rabbit Polyclonal Antibody
Catalog # 10162-120
Supplier:  Genetex
Anti-SHANK3 Rabbit Polyclonal Antibody
Catalog # 10162-120
Supplier:  Genetex
Supplier Number:  GTX60153
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Sh3 And Multiple Ankyrin Repeat Domains 3
  • Antigen Symbol:
    SHANK3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Size:
    100 μg
  • Environmentally Preferable:
  • Gene ID:
    85358
  • Antigen Synonyms:
    PROSAP2|SCZD15|DEL22q13.3|PSAP2|SPANK-2
  • Storage Buffer:
    100ug/ml BSA, 50% glycerol and 0.09% sodium azide.
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at -20oC or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    KLH conjugated synthetic peptide derived from human SHANK3
  • Purification:
    Purified by Protein A and peptide affinity chromatography.
  • Cat. No.:
    10162-120
  • Supplier no.:
    GTX60153

Specifications

About this item

Rabbit Polyclonal antibody to SHANK3

Type: Primary
Antigen: SHANK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat