ContactUsLinkComponent
MTS-32 TD Multiple Tube Sequential Sampler, Restek
MTS-32 TD Multiple Tube Sequential Sampler, Restek
Catalog # 10862-932
Supplier:  Restek
undefined
MTS-32 TD Multiple Tube Sequential Sampler, Restek
Catalog # 10862-932
Supplier:  Restek
Supplier Number:  26464
Specifications
  • Description:
    Multiple TD Tube Sequential Sampler, Markes MTS-32, Requires ACTI-VOC with battery & charger, or power supply, Holds up to 32 thermal desorption tubes
  • Environmentally Preferable:
  • Cat. No.:
    10862-932
  • Supplier no.:
    26464
Specifications
About this item
The MTS-32 TD Multiple Tube Sequential Sampler allows sequential sampling of up to 32 thermal desorption tubes (3.5" each).
  • Worry-free, independent operation lets you get more done
  • Simply set up the unit in the field for extended sampling
  • Retrieve it at the end of the sampling period
  • Reliably collect accurate samples
  • Diffusion-locking technology prevents sample losses or the ingress of contaminants
  • Easy to use and portable
  • Use almost anywhere
  • Sample pollutants in ambient air or continuously monitor industrial emissions