ContactUsLinkComponent
Compound Pendulum
  470220-610
 :  Avantor
undefined
Compound Pendulum
  470220-610
 :  Avantor
  • Description:
    Compound Pendulum
  • Environmentally Preferable:
  • Cat. No.:
    470220-610
  • Supplier no.:
    SP394
The compound pendulum consists of a steel bar (1 Meter long) drilled at 50mm intervals, removable knife edges and a support bracket for mounting on walls.
  • Sturdy Design
  • Repeatable results