Specifications
- Description:STERIL-PEEL Heat Seal Pouches
- Environmentally Preferable:
- Cat. No.:89201-554
- W×L:19.1 cm x 33 m (7.5” x 13’)
- Supplier no.:221633
Specifications
About this item
All STERIL-PEEL™ products are manufactured from medical grade bleached kraft paper
STERIL-PEEL™ products are printed with dual indicators which change color in either steam or ethylene oxide. The steam indicator ink will only change in the presence of all necessary parameters (saturated steam, temperature, and time).
Hydrogen peroxide, rather than chlorine, is used as the bleaching agent. They are made from virgin pulp, ensuring optimum consistency of the paper sheet, providing superior barrier properties.