ContactUsLinkComponent
McFarland Standards Kit, Biomerieux
McFarland Standards Kit, Biomerieux
  95060-984
 :  
McFarland Standards Kit, Biomerieux
  95060-984
 :  70900
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    McFarland Standards Kit
  • Environmentally Preferable:
  • Cat. No.:
    95060-984
  • Supplier no.:
    70900
McFarland standards are used as a reference for adjusting the turbidity of bacterial suspensions so that the number of bacteria will be within a given range to standardize microbial testing.
  • Stable suspensions
  • Allows for visual comparison with bacterial suspensions
Standards are economical and easy to use, with straightforward comparison with bacterial suspensions.