Single-Stage Ultra-High Purity Stainless Steel Gas Regulators with CGA Fittings, Restek
Supplier: Restek
Certificates
About this item
UHP stainless steel regulators are the standard for ultra-high-purity and corrosion-resistant pressure regulation.
- Identical gas purity protection as with dual-stage gas regulators
- Use when there is secondary pressure regulation downstream
- Also available with international fittings
All regulators are rated to 3000 psig maximum inlet pressure.
Specifications
- Catalog No:
- 10855-180
- 10855-622
- 10855-370
- Outlet Gauge:
- 30", 0 to 200 psig (0 – 1379 kPa)
- 30", 0 to 200 psig (0 – 1379 kPa)
- 30", 0 to 200 psig (0 – 1379 kPa)