ContactUsLinkComponent
Hydrogen Flashback Arrestor, Restek
  10855-640
 :  Restek
undefined
Hydrogen Flashback Arrestor, Restek
  10855-640
 :  Restek
 :  21334
  • Description:
    Hydrogen Flashback Arrestor
  • Material:
    Brass Body
  • Environmentally Preferable:
  • Cat. No.:
    10855-640
  • Includes:
    1/4" female NPT
  • Supplier no.:
    21334
Valve cuts off gas supply in the event of a flashback.
  • Can be reset and reused after flashback has occurred
  • Brass body
  • Polyethylene temperature sensor
 : Meet safety requirements of BS 6158, EN 730, ISO 5175, AS 4603 and are UL Listed 23Y5.