To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Control
- Antigen Symbol:IgG2A
- Host:Rat
- Isotype:IgG2a
- Reactivity:
- Size:0.05 mg
- Environmentally Preferable:
- Form:Lyophilized
- Antigen Synonyms:Ig gamma-2A chain C region, Igg-2a|Igg-2a
- Antibody Fragment:Fc
- Purity:> 95% as determined by reducing SDS-PAGE
- Storage Buffer:Lyophilized from a 0.2 um filtered solution of 20mM Tris, pH 8.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Molecular Weight:35.2 kD
- Storage Temperature:Lyophilized protein should be stored at -20°C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7°C for 2-7 days. Aliquots of reconstituted samples are stable at -20°C for 3 months.
- Cat. No.:75791-936
- Supplier no.:92-687
Specifications
About this item
Immunoglobulin G (IgG) is a type of antibody composed of four peptide chains—two identical heavy chains and two identical light chains arranged in a Y-shape typical of antibody monomers.There are four IgG subclasses (IgG1, 2, 3, and 4) in humans, named in order of their abundance in serum. The IgG2a isotype was able to interact very efficiently with FcgammaR, and share approximately 64-78% amino acid sequence identity with IgG1 and IgG2b in the heavy chain constant region domains, CH1, CH2 and CH3, which cause the diffrence in the interaction with complement, NK and mast cells, induction or inhibition by specific cytokines, and ability to cross the placenta.
- Endotoxin content: <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
This recombinant protein can be used for biological assays. For research use only.