To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Environmentally Preferable:
- Tag sequence:N-6 His tag
- Storage Conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme Name:Glycine N-Methyltransferase
- Enzyme Synonyms:Glycine N-Methyltransferase, GNMT
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular Weight:34.9 kD
- Sequence:Met1-Asg294
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75788-672
- Supplier no.:91-999
Glycine N-Methyltransferase (GNMT) is a tetrameric cytosolic protein
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
GNMT catalyzes the synthesis of N-methylglycine from glycine using S-adenosylmethionine (AdoMet) as the methyl donor. It can affects DNA methylation by regulating the ratio of S-adenosylmethionine to S-adenosylhomocystine, playing an important role in maintaining normal AdoMet levels. GNMT is highly expressed in liver. As a major folate-binding protein, GNMT takes part in the detoxification pathway. Defects in GNMT are the cause of hypermethioninemia. the patients with this deficiency are mild hepatomegaly and chronic elevation of serum transaminases.