Order Entry
United States
ContactUsLinkComponent
Human Recombinant alpha-Lactalbumin (from Cells)
Human Recombinant alpha-Lactalbumin (from Cells)
Catalog # 75790-476
Supplier:  Avantor
CAS Number:  
Human Recombinant alpha-Lactalbumin (from Cells)
Catalog # 75790-476
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Environmentally Preferable:
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    LYZL7|LALBA|Lactose synthase B protein|Lysozyme-like protein 7
  • Protein/Peptide Name:
    alpha-Lactalbumin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    15.1 kD
  • Sequence:
    Lys20-Leu142
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM Tris,150mM NaCl,PH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75790-476
  • Supplier no.:
    91-920

Specifications

About this item

Alpha-lactalbumin(LALBA) is a secreted protein that is a member of the glycosyl hydrolase 22 family.It is expressed in mammary gland in milk. It is the regulatory subunit of lactose synthase.It changes the substrate specificity of galactosyltransferase in the mammary gland making glucose a good acceptor substrate for this enzyme.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.