To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:glutathione peroxidase 1
- Antigen Symbol:GPX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:100 ug
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P07203
- Antigen Synonyms:GPX1
- Storage Buffer:Liquid. 0.2um-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage Temperature:Stable for at least 6 months after receipt when stored at –20 °C.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Recombinant human GPX1.
- Cat. No.:75793-440
- Supplier no.:36-306
Specifications
About this item
GPX1 is an antioxidant enzyme that is down-regulated in cancer cells. Protects the hemoglobin in erythrocytes from oxidative breakdown. Its main role is the prevention of cancer initiation by ROS-mediated DNA damage.
ELISA: (direct and indirect: 1:2,000-1:5,000). Western Blot: (1:2,000-1:5,000 using ECL; suggested blocking and dilution buffer is PBST containing 0.05% Tween 20 and 5% skim milk; suggested incubation time is 1 hour at room temperature). Optimal conditions must be determined individually for each application.
Type: Primary
Antigen: GPX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human