ContactUsLinkComponent
Liquid Nitrogen Supply Tanks, Thermo Scientific
Liquid Nitrogen Supply Tanks, Thermo Scientific
Certificates
Liquid Nitrogen Supply Tanks, Thermo Scientific
Specifications
  • Catalog No:
  • 97049-692
  • 101623-292
  • 97049-652
  • 101623-264
  • 97049-654
  • Capacity:
  • 180 L
  • 180 L
  • 230 L
  • 230 L
  • 50 L
  • Ext.Ø:
  • 50.8 cm (20")
  • 51 cm (20")
  • 66 cm (26")
  • 66 cm (26")
  • 40.6 cm (16")
  • Overall Height:
  • 161.3 cm (63.5")
  • 161.3 cm (63.5")
  • 139.2 cm (54.75")
  • 139.2 cm (54.75")
  • 90.2 cm (35.5")