BioBall®, MultiShot 550, bioMerieux
Supplier: bioMerieux
Certificates
About this item
BioBall® MultiShot 550 is ideal for testing the efficacy of antimicrobial preservation.
- Unprecedented accuracy for quantitative microbiological quality control
- BioBall® is an accredited reference material under ISO Guide 34 standard
- Easy to use-no preparation or pre-incbation
Offers unparalleled precision, ease of use and is ideal for industrial applications requiring a known high level of inoculum.
BioBall® contains a precise number of microorganisms in a water soluble ball delivering unprecedented precision and accuracy for Quantitative Microbiological Quality Control.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together