di-Potassium oxalate monohydrate 99+% for analysis
Supplier: Avantor
Certificates
About this item
Synonyms: Oxalic acid potassium salt monohydrate, Potassium oxalate hydrate, Potassium oxalate monohydrate, Potassium ethanedioate monohydrate
CAS Number:6487-48-5
:
Formula:(COOK)₂·H₂O
MDL Number:MFCD00150033
Molecular Weight:184.23 g/mol
Melting Pt:356
Merck Index:12,07820
EINECS:209-506-8
UN:2811
:GHS07
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystals or flakes
- Infrared spectrum:Conforms
- pH:7 to 8.5 (5 % at 20 °C)
- Titration with HClO4:≥99.0 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤100 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤20 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_AC20771
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together