Appearance: Cream to brown Powder
: Cyclohexanehexone octahydrate, Triquinoyl octahydrate, Cyclohexane-1,2,3,4,5,6-hexone octahydrate
- :7255-28-9
- :C₆O₆·8H₂O
- :MFCD00149074
- :312.18 g/mol
- :90…93 °C
- :Ambient
- :2811
- :6.1,III
Appearance: Cream to brown Powder
: Cyclohexanehexone octahydrate, Triquinoyl octahydrate, Cyclohexane-1,2,3,4,5,6-hexone octahydrate
- :7255-28-9
- :C₆O₆·8H₂O
- :MFCD00149074
- :312.18 g/mol
- :90…93 °C
- :Ambient
- :2811
- :6.1,III
Documents
ghs_AC15628
SDS
Product Family Options
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



