To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Fluorescein Isothiocyanate
- Antigen Symbol:FITC
- Clonality:Monoclonal
- Conjugation:Alexa Fluor® 488
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Size:500 μg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:IgG Frac
- Form:Freeze-dried powder
- Storage Buffer:0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, 15mg/mL BSA, 0.05% Azide
- Storage Temperature:+4 °C
- Concentration:1.4 - 1.8 mg/mL
- Shipping Temperature:ambient
- Purification:IgG Frac
- Cat. No.:102647-062
- Packaging:Glass ampoule
- Supplier no.:200-542-037
Specifications
About this item
IgG fraction Monoclonal Mouse Anti-Fluorescein may be used either as direct conjugates, or for more sensitivity, they can be used unconjugated followed by a conjugated anti-mouse IgG (H+L) for signal enhancement
Type:
Antigen: FITC
Clonality: Monoclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: