Workrite® FR Nomex® Men's Knit Cuff Flame Resistant Lab Coat
Supplier: Bulwark Protection
Certificates
About this item
Nomex® Men's Knit Cuff Flame Resistant Lab Coats feature a protective thigh length design, with a traditional collar and knit cuffs
- Protective thigh length design
- Concealed snap-front closures
- Side pass-through access to inner clothing
- Four pockets
- UL Certified to NFPA 2112
These coats are equipped with concealed snap front closures as well as side pass-through access to inner clothing. Each coat has four pockets.
Certifications: UL-Certified to NFPA 2112.
Ordering information: Lab coats 10748-100, 10748-096, 10748-092, 10748-104, 10748-108, 10748-120, 10748-124, and 10748-112 are extra-long versions of the listed size.
Specifications
- Catalog No:
- 10748-106
- 10748-102
- 10748-114
- 10748-110
- 10748-098
- 10748-122
- 10748-094
- 10748-118
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- Fabric:
- Nomex® Men's knit cuff
- Nomex® Men's knit cuff
- Nomex® Men's knit cuff
- Nomex® Men's knit cuff
- Nomex® Men's knit cuff
- Nomex® Men's knit cuff
- Nomex® Men's knit cuff
- Nomex® Men's knit cuff