Order Entry
United States
ContactUsLinkComponent
Fetal Bovine Serum (FBS) LifeFactor, Lifeline Cell Technology
Fetal Bovine Serum (FBS) LifeFactor, Lifeline Cell Technology
Catalog # 102970-742
CAS Number:  
Fetal Bovine Serum (FBS) LifeFactor, Lifeline Cell Technology
Catalog # 102970-742
Supplier Number:  LS-1028
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
  • Species:
  • Description:
    FBS Fetal Bovine Serum LifeFactor
  • Size:
    25 mL
  • Environmentally Preferable:
  • Cat. No.:
    102970-742
  • Supplier no.:
    LS-1028

Specifications

About this item

Fetal Bovine Serum is used with human cells, fibroblasts and pre-adipocytes, and basal medium.

  • Used with human endothelial cells
  • Used with human smooth muscle cells
  • Used with basal medium
  • Used with human peripheral blood cells
  • Used with human pre-adipocytes

Fetal Bovine Serum LifeFactor 25 mL may be used with: Lifeline® VascuLife® basal medium and human smooth muscle cells or human endothelial cells, FibroLife® basal medium and human fibroblasts, RPMI 1640 basal medium and human peripheral blood cells, StemLife PA basal medium and human pre-adipocytes.