To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Retrovirus Quantitation Kit
- Size:20 Assays
- Environmentally Preferable:
- Storage Temperature:4°C
- Sensitivity:10^10 VP/mL
- Cat. No.:102965-358
- Supplier no.:VPK-120
Specifications
About this item
QuickTiter™ Retrovirus Quantitation Kit provides a quick method for measuring the viral nucleic acid content of your retrovirus prep
This assay may be performed either before or after purification of your virus. The kit uses an ultra-fast 45-60 minute procedure to measure the viral nucleic acid content of purified retrovirus or unpurified supernatant. The limit of detection is 10^10 viral particles/mL from 100 µL of retroviral supernatant.