ContactUsLinkComponent
Odyssey® EMSA (Gel Shift) Reagents, LI-COR
Odyssey® EMSA (Gel Shift) Reagents, LI-COR
Catalog # 102673-212
Supplier:  Li-Cor
CAS Number:  
Odyssey® EMSA (Gel Shift) Reagents, LI-COR
Catalog # 102673-212
Supplier:  Li-Cor
Supplier Number:  829-07910
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Odyssey EMSA Buffer Kit
  • Size:
    100 mL
  • Environmentally Preferable:
  • Cat. No.:
    102673-212
  • Supplier no.:
    829-07910
Specifications
About this item
Gel shift assays, or electrophoretic mobility shift assays (EMSA), provide a simple method to study DNA-protein interactions.
  • Can be easily converted to infrared fluorescent assays
  • Conditions are the same as any other EMSA detection method
  • Creates good substrate for protein binding
  • Complete infrared assays in less than two hours
Existing mobility shift assay protocols can be easily converted to infrared fluorescent assays by replacing the existing DNA oligonucleotides with IRDye® infrared dye end-labeled oligonucleotides
A DNA oligonucleotide end-labeled with IRDye 700 infrared dye is a good substrate for protein binding. Using the Odyssey Infrared Imaging System, IRDye infrared dye labeled DNA detection is linear within a 50-fold dilution range from 9.1 fmol to 0.18 fmol. Infrared assays can be completed in less than two hours with no gel transfer or film exposure. The gel doesn’t even have to be removed from the glass plates for imaging. Plus, the gel can be placed back into the electrophoresis unit and run longer, if necessary.
Binding and electrophoresis conditions are the same as any other EMSA detection method. You can easily adapt existing protocols by replacing radiolabeled oligos with IRDye® 700 and IRDye 800 end-labeled oligos.
Ordering information: IRDye 700 infrared dye-labeled EMSA oligonucleotides are supplied as 25 µL of 50 nM double-stranded DNA.