Order Entry
United States
ContactUsLinkComponent
Total Carbohydrate Assay Kit, Cell Biolabs
Total Carbohydrate Assay Kit, Cell Biolabs
  102966-256
 :  
undefined
Total Carbohydrate Assay Kit, Cell Biolabs
  102966-256
 :  STA-682
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    Total Carbohydrate Assay Kit
  • Size:
    100 Assays
  • Environmentally Preferable:
  • Storage Temperature:
    Room Temperature and –4°C
  • Sensitivity:
    62.5 µM
  • Cat. No.:
    102966-256
  • Method:
    Colorimetric
  • Supplier no.:
    STA-682

 

 

Cell Biolabs' Total Carbohydrate Assay detects total carbohydrates within samples based on the phenol-sulfuric acid method

Carbohydrates are hydrolyzed to furfural and furfural derivates upon addition of sulfuric acid, which then react with the Developing Solution to form a chromagen that is detected at 490nm. Total carbohydrate levels in an unknown sample are calculated based on a glucose standard curve.