Order Entry
United States
ContactUsLinkComponent
Anti-IgG Donkey Polyclonal Antibody (IRDye® 680RD)
Anti-IgG Donkey Polyclonal Antibody (IRDye® 680RD)
Catalog # 102971-026
Supplier:  Li-Cor
Anti-IgG Donkey Polyclonal Antibody (IRDye® 680RD)
Catalog # 102971-026
Supplier:  Li-Cor
Supplier Number:  925-68074
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    IRDye® 680RD
  • Host:
    Donkey
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Goat
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Cross Adsorption:
    Highly Cross-adsorbed
  • Form:
    Lyophilized
  • Storage Buffer:
    PBS, pH 7.4. Contains 10 mg/mL BSA (free of IgG and protease) as stabilizer and 0.1% sodium azide as preservative, after reconstitution
  • Storage Temperature:
    Protect from light. Store at 4 °C prior to reconstitution
  • Tested Applications:
    On-Cell Western Assay, Protein Array, Microscopy, 2D Gel Detection, Tissue Section Imaging, Small Animal Imaging = Yes
  • Cat. No.:
    102971-026
  • Supplier no.:
    925-68074

Specifications

About this item

Isolation of specific antibodies was accomplished by affinity chromatography using pooled IgG covalently linked to agarose. Based on ELISA, this antibody reacts with the heavy and light chains of goat IgG and with sheep IgG. This antibody was tested by Dot Blot and/or solid-phase adsorbed for minimal cross-reactivity with human, mouse, rabbit, rat, chicken, guinea pig, hamster, swine, and horse serum proteins, but may cross-react with immunoglobulins from other species. The conjugate has been specifically tested and qualified for Western blot and In-Cell Western™ assay applications. Highly Recommended for: Western blot, In-Cell Western Assay, On-Cell Western Assay, protein array, immunohistochemistry, microsciopy, 2D gel detection, tissue section imaging, and small animal imaging.

Western blot: 1:15000; In-Cell Western Assay: 1:800

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: IRDye 680RD
Epitope:
Host: Donkey
Isotype:
Reactivity: Goat