Anti-RCAN3 Rabbit Polyclonal Antibody
Catalog # 89512-676
Supplier: Abgent
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Calcipressin 3
- Antigen Symbol:RCAN3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Environmentally Preferable:
- Epitope:C-Terminal
- Gene ID:11123
- Antigen Synonyms:Down syndrome candidate region 1 like protein 2|hRCN3|Down syndrome candidate region 1 like 2|Calcipressin-3|Myocyte-enriched calcineurin-interacting protein 3|RCN3|Down syndrome critical region gene 1 like 2|RCAN3_HUMAN|DSCR1L2|RCAN3|Myocyte enriched calcineurin interacting protein 3|RCAN family member 3|Regulator of calcineurin 3.|regulator of calcineurin 3|Down syndrome candidate region 1-like protein 2|MCIP3
- Amino Acid Number:194 to 222
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:27.492 kDa
- Sequence:EETKNPKQKIAQTRC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.35mg/ml
- Shipping Temperature:4 °C
- Immunogen:This RCAN3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 194-222 amino acids from the C-terminal region of human RCAN3.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89512-676
- Supplier no.:AP14397B
Specifications
About this item
Western Blot: 1:1000, Immunhistochemsitry: 1:10~50,
Type: Primary
Antigen: RCAN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human